Sterilin™ Weighing Boats
Sterilin™ Weighing Boats
Actual product may vary
Sterilin™ Weighing Boats
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ColorTypeShape
WB30338100 mLBlackAntistatic Weighing DishDiamond
FKS118647125 mLWhiteAntistatic Weighing DishDiamond
FKS118447127 mLBlackAntistatic Weighing DishSquare
FKS113338147 mLWhiteStandardSquare
FKS1180471230 mLWhiteStandardDiamond
FKS11804722100 mLWhiteAntistatic Weighing DishSquare
FKS11824712100 mLWhiteStandardSquare
FKS11834712250 mLWhiteStandardSquare
Catalog number WB30338
Price (SGD)
-
Capacity (Metric):
100 mL
Color:
Black
Type:
Antistatic Weighing Dish
Shape:
Diamond
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners. All sizes packed 250 per pack, 500 per case.
  • Manufactured from high impact polystyrene
  • Hydrophobic surface ensures no absorption of water from atmosphere or sample
  • Available in black or white to ensure suitability for different colored sample materials
  • Anti-static range alleviates issues with powder hang-up
  • Flat base for a secure, firm rest on the balance pan
  • Supplied nonsterile
Specifications
Bottom ShapeFlat
Capacity (Metric)100 mL
ColorBlack
MaterialPolystyrene
ShapeDiamond
SterilityNon-sterile
TypeAntistatic Weighing Dish
Length (Metric)120 mm
Width (Metric)100 mm
Unit SizeEA