Deep Vacuum Oil Pumps for Vacuum Concentrators
Deep Vacuum Oil Pumps for Vacuum Concentrators
Actual product may vary
Deep Vacuum Oil Pumps for Vacuum Concentrators
Achieve maximum solute recovery with the Thermo Scientific™Deep Vacuum Oil Pump.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDisplacementVoltage
VLP200-230195L/min. (at 60Hz); 162L/min. (at 50Hz)230 V
VLP120-230
also known as AVL
116L/min. (at 60Hz); 97L/min. (at 50Hz)230 V
Catalog number VLP200-230
Price (SGD)
8,695.00
EA
Add to cart
Displacement:
195L/min. (at 60Hz); 162L/min. (at 50Hz)
Voltage:
230 V
Price (SGD)
8,695.00
EA
Add to cart
Achieve maximum solute recovery with the Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators, designed to work with all Thermo Scientific™ SpeedVac™ concentrating and freeze drying systems.
Features:
  • Three-position gas ballast control optimizes pump performance to suit all applications
  • Large, sideview sight glass allows for convenient checking of proper oil levels at a glance
All Rotary Vane Oil Pumps come standard with:
  • Muffler (OME190 Oil Mist Eliminator)
  • Two Oil Filters (MF190)
  • Quick Drain Kit
Specifications
DescriptionVLP200; 230V 50Hz
TypeDeep Vacuum Oil Pump
Displacement195L/min. (at 60Hz); 162L/min. (at 50Hz)
For Use With (Equipment)Vacuum Concentrator
Oil Capacity0.75 L
Ultimate Vacuum Level1.5 mtorr/1.95 x10³mbar
Vacuum Inlet Fitting12.7 OD
Width (English)6 in.
Width (Metric)160 mm
Height (English)11 in.
Height (Metric)260 mm
Depth (English)19 in.
Depth (Metric)470 mm
Weight (English)57 lb.
Weight (Metric)26 kg
Electrical Requirements230 V 50 Hz
Voltage230 V
Frequency50 Hz
Unit SizeEA