4000 Protective Coverall
Actual product may vary
4000 Protective Coverall
4000 Protective Coverall
4000 Protective Coverall
4000 Protective Coverall
4000 Protective Coverall
4000 Protective Coverall
4000 Protective Coverall
ULTITEC
4000 Protective Coverall
Designed for hazardous conditions with Type 3, Type 4, Type 5 and EN14126 certification.
Have Questions?
Catalog NumberSize
UTCDD610-XLX-Large
Catalog number UTCDD610-XL
Price (SGD)
660.00
Special offer
Ends: 06-Sep-2026
803.00
Save 143.00 (18%)
Case
Size:
X-Large
Price (SGD)
660.00
Special offer
Ends: 06-Sep-2026
803.00
Save 143.00 (18%)
Case
  • Chemical and Liquid Jet Resistant Protective Clothing.
  • An Outstanding Barrier Against Biological Hazards.
  • Proven Highest Protection of Fabric Performance Against Blood or Body Fluids.
  • Highest Protection of Fabric Performance Against Blood-borne Pathogens.
Specifications
Product TypeChemical Protective Clothing
SizeX-Large
GenderUnisex
MaterialFabric
ColorYellow
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeAttached, Open Face
Seam TypeSealed
SterilityNon-sterile
ResistanceCaustics, Acid
Unit SizeCase