2000 Protective Coverall
Actual product may vary
2000 Protective Coverall
2000 Protective Coverall
2000 Protective Coverall
2000 Protective Coverall
2000 Protective Coverall
ULTITEC
2000 Protective Coverall
Designed to protect wearers against heavy liquid splash and biological hazards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
UTCDD310-XLX-Large
UTCDD310-LLarge
Catalog number UTCDD310-XL
Price (SGD)
330.00
Special offer
Ends: 06-Sep-2026
403.00
Save 73.00 (18%)
Case
Size:
X-Large
Price (SGD)
330.00
Special offer
Ends: 06-Sep-2026
403.00
Save 73.00 (18%)
Case
  • High Moisture Vapor Transmission rate keeps your people cool and comfortable.
  • Well-designed garment provides worker comfort and a lower cost-in-use.
  • Suitable to use in ISO class 6 and above cleanroom with controlled environment.
Specifications
Product TypeChemical Protective Clothing
SizeX-Large
GenderUnisex
MaterialFabric
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeAttached, Open Face
Seam TypeSealed
SterilityNon-sterile
ResistanceLiquid, Chemicals, Dust
Unit SizeCase