1000L Elasticated Protective Coverall
Actual product may vary
1000L Elasticated Protective Coverall
1000L Elasticated Protective Coverall
1000L Elasticated Protective Coverall
1000L Elasticated Protective Coverall
1000L Elasticated Protective Coverall
ULTITEC
1000L Elasticated Protective Coverall
Protective Coverall with elasticated hood, wrist, waist and ankles.
Have Questions?
Catalog NumberSize
UTCDD210-LLarge
Catalog number UTCDD210-L
Price (SGD)
210.00
Special offer
Ends: 31-Dec-2026
253.00
Save 43.00 (17%)
Case
Size:
Large
Price (SGD)
210.00
Special offer
Ends: 31-Dec-2026
253.00
Save 43.00 (17%)
Case
  • High Moisture Vapor Transmission rate keeps your people cool and comfortable.
  • Well-designed garment provides worker comfort and a lower cost-in-use.
  • Antistatic offers protection for workers who are exposed to environments where sparks could cause explosions or ignition.
Specifications
Product TypeProtective Coveralls
SizeLarge
GenderUnisex
MaterialMicroporous Fabric
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeAttached, Open Face
Seam TypeSealed
SterilityNon-sterile
ResistanceDust, Liquid
Unit SizeCase