Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Solvents
▸
Other Solvents
▸
Xylenes
▸
Xylenes (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Xylenes (Certified ACS), Fisher Chemical™
Mixture of ortho, meta, and para isomers and may contain some Ethylbenzene
Have Questions?
Catalog Number
Quantity
Packaging
USFX51
1 L
Glass Bottle
Catalog number
USFX51
Price (SGD)
76.00
Each
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
76.00
Each
Add to cart
Specifications
Chemical Name or Material
Xylenes
Cleanup Symbol
Solvent
Melting Point
-34°C
CAS
1330-20-7
ChemAlert Storage Symbol
Red
View more