▸
Solvents
▸
Tetrahydrofuran
▸
Tetrahydrofuran, Optima™ for HPLC, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Tetrahydrofuran, Optima™ for HPLC, Fisher Chemical™
High purity solvent for HPLC
Have Questions?
Catalog Number
Quantity
Packaging
USFT4271
1 L
Wide Neck Amber Bottle
Catalog number
USFT4271
Price (SGD)
100.00
Each
Add to cart
Quantity:
1 L
Packaging:
Wide Neck Amber Bottle
Request bulk or custom format
Price (SGD)
100.00
Each
Add to cart
Specifications
Chemical Name or Material
Tetrahydrofuran
pH
7 to 8
Cleanup Symbol
Solvent
Melting Point
-108.4°C
Absorbance
1.0 AU max. at 210nm, 0.6 AU max. at 215nm, 0.3 AU max. at 230nm, 0.1 AU max. at 254nm
View more
Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
Also meets ACS specifications
Intended for research use only