Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Silver Nitrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Silver Nitrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
Standardized at 25°C against NIST™ potassium chloride
Have Questions?
Catalog Number
Quantity
Packaging
USFSS72500
500 mL
Glass Bottle
Catalog number
USFSS72500
Price (SGD)
101.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
101.00
Each
Add to cart
Specifications
Chemical Name or Material
Silver Nitrate Solution
Color
Colorless
Name Note
0.1N (N/10)
CAS
7761-88-8
ChemAlert Storage Symbol
Yellow
View more