▸
Bases
▸
Sodium Hydroxide
▸
Sodium Hydroxide Solution (2N/Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide Solution (2N/Certified), Fisher Chemical
A volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
USFSS2641
1 L
Poly Bottle
Catalog number
USFSS2641
Price (SGD)
57.00
Each
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
57.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
pH
12.0
Cleanup Symbol
Caustic
Name Note
2N
Boiling Point
100°C
View more