▸
Bases
▸
Sodium Hydroxide
▸
Sodium Hydroxide Solution (10N/Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide Solution (10N/Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFSS2551
1 L
Poly Bottle
Catalog number
USFSS2551
Price (SGD)
64.00
Each
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
64.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
pH
12.0
Cleanup Symbol
Caustic
Name Note
10N
Boiling Point
>100°C
View more