▸
Buffers and Standards
▸
Electrochemistry Buffers
▸
Buffer Solution, pH 4.00 (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Buffer Solution, pH 4.00 (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFSB98500
500 mL
Poly Bottle
Catalog number
USFSB98500
Price (SGD)
40.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
40.00
Each
Add to cart
Chemical Identifiers
CAS
877-24-7, 7732-18-5, 50-00-0, 67-56-1
Specifications
Chemical Name or Material
Buffer Solution, pH 4.00
CAS
877-24-7
ChemAlert Storage Symbol
Gray
Color
Undesignated
Health Hazard 2
CAUTION!
View more