▸
Acids
▸
Inorganic Acids
▸
Hydrochloric Acid
▸
Hydrochloric Acid Solution, 1N (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid Solution, 1N (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
USFSA481
1 L
Poly Bottle
Catalog number
USFSA481
Price (SGD)
76.00
Each
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
76.00
Each
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid Solution, 1N
pH
1.1
Cleanup Symbol
Acid
CAS
7647-01-0
ChemAlert Storage Symbol