Check Order Status
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica gel Grade 923 conforms to the specifications of ASTM D1319 and ASTM D2549.
Have Questions?
Catalog Number
Quantity
Packaging
USFS679500
500 g
Poly Bottle
Catalog number
USFS679500
Price (SGD)
855.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
855.00
EA
Add to cart
Specifications
Boiling Point
2230°C
Chemical Name or Material
Silica Gel Desiccant
Color
White
Melting Point
1710°C
Physical Form
Solid
View more
Sold as a case of 3 boxes - each box contains 24 x 10 g pouches
Suitable for use in Fluorescent Indicator Adsorption (FIA) test
Conforms to the specifications of ASTM D1319 and ASTM D2549
Surface area within the range of 430 - 530 m2/g
This grade of silica gel is also available in other sizes:
500 g (S679500)
2.5 kg (S679212)
25 g (S67925)