Storage Vessel with High Vacuum Valve
Actual product may vary
Fisher Chemical
Storage Vessel with High Vacuum Valve
Storage Vessel features a #15 O-ring joint on the sidearm and a high vacuum valve plug to control gas and/or liquid flow into the vessel
Have Questions?
Catalog NumberCapacity (Metric)
USFS490100100 mL
Catalog number USFS490100
Price (SGD)
69.00
Each
Capacity (Metric):
100 mL
Price (SGD)
69.00
Each
Specifications
ColorClear
Capacity (Metric)100 mL
MaterialBorosilicate Glass
For Use With (Application)Organic Synthesis
Height (Metric)235 mm
Unit SizeEach