▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFS415212
2.5 kg
Glass Bottle
Catalog number
USFS415212
Price (SGD)
142.00
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
142.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Sulfate Anhydrous
Melting Point
884°C
CAS
7757-82-6
ChemAlert Storage Symbol
Gray
Color
White
View more