Check Order Status
Sodium meta-Silicate Nonahydrate (Crystalline/Certified), Fisher Chemical
Element Search
Structure Search
Sodium meta-Silicate Nonahydrate (Crystalline/Certified), Fisher Chemical
Have Questions?
Catalog number
USFS408500
Price (SGD)
-
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium meta-Silicate Nonahydrate
Melting Point
72°C
CAS
13517-24-3
ChemAlert Storage Symbol
White
Color
White
View more