▸
Bases
▸
Sodium Hydroxide
▸
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFS318500
500 g
Poly Bottle
Catalog number
USFS318500
Price (SGD)
59.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
59.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide
pH
14
Cleanup Symbol
Caustic
Color
White
Boiling Point
1390°C
View more