Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™
Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™
Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™
Fisher Chemical
Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
USFS279500500 gPoly Bottle
Catalog number USFS279500
Price (SGD)
67.00
Each
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
67.00
Each
Specifications
Chemical Name or MaterialSodium Citrate Dihydrate
CAS6132-04-3
Health Hazard 2CAUTION!
Decomposition Information>230°C
Health Hazard 3Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
View more