Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Potassium chloride for calomel cells
Have Questions?
Catalog Number
Quantity
Packaging
USFP217500
500 g
Poly Bottle
Catalog number
USFP217500
Price (SGD)
73.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
73.00
Each
Add to cart
Specifications
Boiling Point
1420°C
Chemical Name or Material
Potassium Chloride
Melting Point
770°C
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
View more