▸
Organic compounds
▸
Organic Polymers
▸
Methyl Cellulose (Powder/USP/FCC), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Methyl Cellulose (Powder/USP/FCC), Fisher Chemical
4000 centipoise
Have Questions?
Catalog Number
Quantity
Packaging
USFM352500
500 g
Glass Bottle
Catalog number
USFM352500
Price (SGD)
246.00
Each
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
246.00
Each
Add to cart
Specifications
Chemical Name or Material
Methyl Cellulose
Color
White
pH
5.0 - 8.0
CAS
9004-67-5
ChemAlert Storage Symbol
Gray
View more