▸
Solvents
▸
Hexane
▸
Hexanes (HPLC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hexanes (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Have Questions?
Change view
Quantity:
4 L
Packaging:
Glass Bottle
Catalog number
USFH3024
Price (SGD)
116.00
Bottle
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
116.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Hexanes
Cleanup Symbol
Solvent
Melting Point
-95°C
Absorbance
1.00 AU max. at 195nm, 0.25 AU max. at 210nm, 0.08 AU max. at 220nm, 0.014 AU max. at 254nm
CAS
92112-69-1
View more