Hexamethylenetetramine (Crystalline/USP), Fisher Chemical
Hexamethylenetetramine (Crystalline/USP), Fisher Chemical
Hexamethylenetetramine (Crystalline/USP), Fisher Chemical
Fisher Chemical
Hexamethylenetetramine (Crystalline/USP), Fisher Chemical
Have Questions?
Catalog NumberQuantityPackaging
USFH290500500 gGlass Bottle
Catalog number USFH290500
Price (SGD)
459.00
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
459.00
Each
Specifications
Chemical Name or MaterialHexamethylenetetramine
CAS100-97-0
ChemAlert Storage SymbolRed
Health Hazard 1Signal Word: WARNING!
Health Hazard 3Emergency Overview
Flammable solid. May cause an allergic skin reaction.
NFPA
Health: 2
Flammability:
View more