▸
Biochemical Reagents
▸
Carbohydrates
▸
Sugar alcohols
▸
Glycerol (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Glycerol (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFG331
1 L
HDPE Bottle
Catalog number
USFG331
Price (SGD)
66.00
Each
Add to cart
Quantity:
1 L
Packaging:
HDPE Bottle
Request bulk or custom format
Price (SGD)
66.00
Each
Add to cart
Specifications
Chemical Name or Material
Glycerol
CAS
56-81-5
ChemAlert Storage Symbol
Gray
Health Hazard 2
WARNING!
Decomposition Information
>290°C
View more