HardyCHROM™ Staphylococcus Aureus Chromogenic Medium
Actual product may vary
Fisher Chemical
HardyCHROM™ Staphylococcus Aureus Chromogenic Medium
HardyCHROM™ Staph aureus is a chromogenic medium recommended for the isolation, differentiation, and enumeration of Staphylococcus aureus by colony color
Have Questions?
Catalog number USFG311
Price (SGD)
75.00
Each
Price (SGD)
75.00
Each
  • HardyCHROM™ Staph aureus allows for the rapid and reliable detection of S. aureus from both clinical and food specimens within 24 hours
  • Selective agents inhibit the growth of gram-negative organisms, yeast, and some gram-positive cocci
Specifications
AppearanceTranslucent
ColorLight Amber
DetectsStaphylococcus aureus
FormPrepared Medium
Manufacturing OriginUnited States
Media TypeChromogenic Medium
Product TypeAgar Plate
Target Organism ClassGram-positive Cocci
Test TypeMicrobial Detection
Fill Volume18 mL
For Use With (Application)Isolation, Differentiation, Enumeration of Staphylococcus aureus
Incubation Conditions35°C
Incubation Time24 hr.
Packaging TypeBox
Dimensions (Dia. x H)100 x 15 mm
Height (Metric)100 mm
Diameter (Metric)15 mm
SpeciesStaphylococcus aureus, Staphylococcus species
Unit SizeEach