DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
Fisher Chemical
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF processed to high purity for accurate and repeatable determination of trace levels of residual solvents.
Have Questions?
Catalog NumberQuantityPackaging
USFD13311 LGlass Bottle
Catalog number USFD1331
Price (SGD)
195.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
195.00
Each
Specifications
Chemical Name or MaterialDMF, Dimethylformamide
Absorbance0.300 AU Maximum (275nm) , 0.050 AU Maximum (300nm), 0.005 AU Maximum (350nm), 0.005 AU Maximum (400nm)
CAS68-12-2
ColorColorless
Recommended StorageRoom temperature, packaged under inert atmosphere.
View more
  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • This product is UV Absorbance Tested to ensure low organic contamination
  • Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC