Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
HPLC mobile phase for analytical and preparative separations
Have Questions?
Catalog NumberQuantityPackaging
USFA99844 LAmber Glass Bottle
Catalog number USFA9984
Price (SGD)
185.00
Each
Quantity:
4 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (SGD)
185.00
Each
Specifications
Chemical Name or MaterialAcetonitrile
Cleanup SymbolSolvent
Melting Point-46°C
Absorbance0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test
CAS75-05-8
View more