Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Acids
▸
Organic Acids
▸
Citric Acid
▸
Citric Acid, Anhydrous (Crystalline/USP), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Citric Acid, Anhydrous (Crystalline/USP), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFA953
3 kg
Poly Bottle
Catalog number
USFA953
Price (SGD)
177.00
Each
Add to cart
Quantity:
3 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
177.00
Each
Add to cart
Specifications
Chemical Name or Material
Citric Acid, Anhydrous
CAS
77-92-9
ChemAlert Storage Symbol
Gray
Melting Point
153°C
Boiling Point
175°C
View more