Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Solvents
▸
Acetone
▸
Acetone (HPLC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetone (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Have Questions?
Catalog Number
Quantity
Packaging
USFA9494
4 L
EcoSafPak™ Amber Glass Bottle
Catalog number
USFA9494
Price (SGD)
78.00
Each
Add to cart
Quantity:
4 L
Packaging:
EcoSafPak™ Amber Glass Bottle
Request bulk or custom format
Price (SGD)
78.00
Each
Add to cart
Specifications
Chemical Name or Material
Acetone
pH
7
Cleanup Symbol
Solvent
Melting Point
-95°C
Absorbance
0.01 AU max. at 400nm, 0.02 AU max. at 350nm, 0.06 AU max. at 340nm, 1.00 AU max. at 330nm
View more