Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Fisher Chemical
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Packaging:
View all
Wide Neck Amber Bottle
Catalog number USFA9291
Price (SGD)
97.00
Each
Quantity:
1 L
Packaging:
Wide Neck Amber Bottle
Request bulk or custom format
Price (SGD)
97.00
Each
Specifications
Chemical Name or MaterialAcetone
pH7
Cleanup SymbolSolvent
Melting Point-95°C
Absorbance1.00 AU max. 330nm, 0.06 AU max. at 340nm, 0.010 AU max. 350nm, 0.01 AU max. (350 to 400nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 300-400nm range, to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only