▸
Salts and Inorganics
▸
Inorganic Salts
▸
Ammonium Salts
▸
Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
USFA639500
500 g
Poly Bottle
Catalog number
USFA639500
Price (SGD)
87.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
87.00
Each
Add to cart
Specifications
Chemical Name or Material
Ammonium Acetate
Melting Point
114°C
CAS
631-61-8
ChemAlert Storage Symbol
Gray
Color
White
View more