Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Fisher Chemical
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
FEP Bottle
Catalog number USFA467500
Price (SGD)
1,224.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
FEP Bottle
Request bulk or custom format
Price (SGD)
1,224.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNitric Acid
pH1
Cleanup SymbolAcid
ColorColorless Yellow
CAS7697-37-2
View more
Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially manufactured and cleaned FEP fluoropolymer bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.