▸
Solvents
▸
Isopropanols
▸
Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™
High purity mobile phase for HPLC and extraction solvent
Have Questions?
Catalog Number
Quantity
Packaging
USFA4644
4 L
Amber Soda Lime Glass
Catalog number
USFA4644
Price (SGD)
126.00
Bottle
Add to cart
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Price (SGD)
126.00
Bottle
Add to cart
Specifications
Chemical Name or Material
2-Propanol
pH
7
Cleanup Symbol
Solvent
Melting Point
-89.5°C
Absorbance
0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
View more
High purity mobile phase for HPLC and extraction solvent
Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
Also meets ACS specifications
Intended for research use only