Ethanol, Anhydrous (Histological), Fisher Chemical™
Ethanol, Anhydrous (Histological), Fisher Chemical™
Ethanol, Anhydrous (Histological), Fisher Chemical™
Ethanol, Anhydrous (Histological), Fisher Chemical™
Ethanol, Anhydrous (Histological), Fisher Chemical™
Fisher Chemical
Ethanol, Anhydrous (Histological), Fisher Chemical™
Used in histology, pathology, and cell biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
4 L
Packaging:
View all
Lined Steel Drum
Catalog number USFA40520
Price (SGD)
313.00
EA
Add to cart
Quantity:
20 L
Packaging:
Lined Steel Drum
Request bulk or custom format
Price (SGD)
313.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthanol, Anhydrous
Melting Point-90°C
CAS64-17-5
ChemAlert Storage SymbolRed
CAS108-10-1
View more