▸
Acids
▸
Inorganic Acids
▸
Sulfuric Acid
▸
Sulfuric Acid (ACS Grade), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sulfuric Acid (ACS Grade), Fisher Chemical™
Approx. Normality is 36
Have Questions?
Change view
Quantity:
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number
USFA300500
Price (SGD)
63.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
63.00
Each
Add to cart
Specifications
Chemical Name or Material
Sulfuric Acid
pH
0.3
Cleanup Symbol
Acid
Color
Colorless to Brown
CAS
7664-93-9
View more