▸
Organic compounds
▸
Organic acids and derivatives
▸
Carboxylic acids and derivatives
▸
Monocarboxylic acids and derivatives
▸
Propionic Acid (Certified ACS), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Propionic Acid (Certified ACS), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
USFA258500
500 mL
Glass Bottle
Catalog number
USFA258500
Price (SGD)
56.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
56.00
Each
Add to cart
Specifications
Chemical Name or Material
Propionic Acid
Cleanup Symbol
Acid
CAS
79-09-4
ChemAlert Storage Symbol
Red
Boiling Range
3°C max. (1 to 95mL)
View more