Check Order Status
Nitric Acid (Certified ACS Plus), Fisher Chemical™
Structure Search
Nitric Acid (Certified ACS Plus), Fisher Chemical™
Nitric Acid material may darken during storage due to photochemical reaction
Have Questions?
Catalog Number
Quantity
Packaging
USFA200500
500 mL
Glass Bottle
Catalog number
USFA200500
Price (SGD)
77.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
77.00
EA
Add to cart
Specifications
Chemical Name or Material
Nitric Acid
pH
-1.0
Cleanup Symbol
Acid
Color
Colorless Yellow
CAS
7697-37-2
View more