Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™
Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™
Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™
Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™
C2HF3O2, CAS Number-76-05-1, acetic acid, trifluoro, cf3cooh, kyselina trifluoroctova, perfluoroacetic acid, trifluoroacetic acid, trifluoroaceticacid, trifluoro acetic acid, trifluoro-acetic acid, trifluoroacetic acid, trifluoroethanoic acid, 100mL, 72 deg.C, CHEBI:45892, Colorless, 114.02g/mol | CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Catalog NumberQuantity
UKFT3258PB05
also known as UKFT/3258/PB05
100 mL
Catalog number UKFT3258PB05
also known as UKFT/3258/PB05
Price (SGD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
pH0.45
CAS76-05-1
Melting Point-15°C
Boiling Point72°C
ColorColorless
View more
C2HF3O2, CAS Number-76-05-1, acetic acid, trifluoro, cf3cooh, kyselina trifluoroctova, perfluoroacetic acid, trifluoroacetic acid, trifluoroaceticacid, trifluoro acetic acid, trifluoro-acetic acid, trifluoroacetic acid, trifluoroethanoic acid, 100mL, 72 deg.C, CHEBI:45892, Colorless, 114.02g/mol