Isopropanol, for HPLC, Fisher Chemical™
Isopropanol, for HPLC, Fisher Chemical™
Isopropanol, for HPLC, Fisher Chemical™
Isopropanol, for HPLC, Fisher Chemical™
Isopropanol, for HPLC, Fisher Chemical™
C3H8O, CAS Number-67-63-0, avantine, dimethylcarbinol, 2-propanol, alkolave, 2-hydroxypropane, hartosol, isopropanol, sec-propyl alcohol, isopropyl alcohol, petrohol, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Catalog number UKFP750717
also known as UKFP/7507/17
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
pH7
Melting Point-89.5°C
CAS67-63-0
ColorColorless
Boiling Point81°C
View more
C3H8O, CAS Number-67-63-0, avantine, dimethylcarbinol, 2-propanol, alkolave, 2-hydroxypropane, hartosol, isopropanol, sec-propyl alcohol, isopropyl alcohol, petrohol, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096