Check Order Status
Petroleum Ether 100-120°C, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Petroleum Ether 100-120°C, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
UKFP156015
also known as UKFP/1560/15
1 L
Amber Glass Bottle
Catalog number
UKFP156015
also known as UKFP/1560/15
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
CAS
64742-49-0
Color
Colorless
Boiling Point
100°C
Quantity
1 L
Packaging
Amber Glass Bottle
View more