Methanol, Certified AR for Analysis, Fisher Chemical™
Methanol, Certified AR for Analysis, Fisher Chemical™
Methanol, Certified AR for Analysis, Fisher Chemical™
Methanol, Certified AR for Analysis, Fisher Chemical™
Methanol, Certified AR for Analysis, Fisher Chemical™
CH4O, CAS Number-67-56-1, wood naphtha, colonial spirit, pyroxylic spirit, methyl hydroxide, methyl alcohol, wood spirit, wood alcohol, methylol, columbian spirit, carbinol, 2.5L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042 | CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Catalog number UKFM400017
also known as UKFM/4000/17
Price (SGD)
-
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Melting Point-98°C
CAS67-56-1
ColorColorless
Boiling Point64.7°C
Quantity2.5 L
View more
CH4O, CAS Number-67-56-1, methyl alcohol, colonial spirit, wood naphtha, wood spirit, wood alcohol, methylol, pyroxylic spirit, methyl hydroxide, columbian spirit, carbinol, 2.5L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042