Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
UKFF1501PB17
also known as UKFF/1501/PB17
2.5 LHDPE plastic bottle
Catalog number UKFF1501PB17
also known as UKFF/1501/PB17
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
PackagingHDPE plastic bottle
CAS50-00-0
Vapor Pressure2mbar at 20°C
Vapor Density> 1.0
Physical FormLiquid
View more
CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C