Check Order Status
Tripod
Tripod
Bochem™ Tripod available with welded feet or screwed feet.
Have Questions?
Catalog Number
Description
Diameter (Metric) Inner
Length (Metric)
Material
TYL5203
Welded Feet
160 mm
240 mm
Stainless Steel (18/10)
Catalog number
TYL5203
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Description:
Welded Feet
Diameter (Metric) Inner:
160 mm
Length (Metric):
240 mm
Material:
Stainless Steel (18/10)
Welded feet on Stainless Steel
Screwed feet on Tempered Cast Iron
Specifications
Material
Stainless Steel (18/10)
Length (Metric)
240 mm
Diameter (Metric) Inner
160 mm
Product Type
Tripod
Description
Welded Feet