Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips
Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips
Actual product may vary
Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips
Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips help deliver samples with a nice uniform bead, cleanly breaking liquid surface tension easily releasing liquid from the distal tip end.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFormat
TTW110RLN-QNon-sterileReload System
QSPTTW110RLN-QNon-sterileReload System
TTW110RL-QNon-sterileReload System
QSPTTW110RL-QNon-sterileReload System
Catalog number TTW110RLN-Q
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Sterility:
Non-sterile
Format:
Reload System
Thermo Scientific™ QSP™ Thin Wall Micro Point 200μl natural pipette tips. Packaged in reload system and non-sterile clean claims are; Rnase/Dnase, DNA, pyrogen and endotoxin free. Universal fit on most pipettes.
  • Thin wall micropoint design decreases surface tension helping liquid bead up cleanly
  • Great for plating
  • Yellow and natural in a variety of packaging options
Specifications
Volume (Metric)200 μL
SterilityNon-sterile
ColorNatural
AutoclavableAutoclavable
Clean ClaimsNon-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
FormatReload System
Packaging4800/Cs.
For Use With (Equipment)Universal Fit
Product LineQSP
Material100% Virgin Polypropylene
Unit SizeEA