Precision™ Shaking Water Baths
Precision™ Shaking Water Baths
Actual product may vary
Precision™ Shaking Water Baths
Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelDescriptionCapacity (Metric)Electrical Requirements
TSSWB15SPrecision SWB 15S15L Shallow Shaking Water Bath15 LGlobal Voltage 100/115 V 200/230 V 50/60 Hz
TSSWB15Precision SWB 1515L Shaking Water Bath15 LGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Catalog number TSSWB15S
Price (SGD)
13,723.00
EA
Add to cart
Model:
Precision SWB 15S
Description:
15L Shallow Shaking Water Bath
Capacity (Metric):
15 L
Electrical Requirements:
Global Voltage 100/115 V 200/230 V 50/60 Hz
Price (SGD)
13,723.00
EA
Add to cart
Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere
  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor
  • Warranty varies by country, please contact us for details
Not for use for IVD/MD in some countries.
Specifications
Chamber Material304 Stainless Steel
Controller TypeDigital
Description15L Shallow Shaking Water Bath
Capacity (Metric)15 L
Certifications/ComplianceUL, CE
Temperature RangeAmbient +5° to 100°C
DisplayMonochrome LCD
Heating Capacity1200 w
IncludesStainless Steel Gable Cover and Rubber Duck
MaterialEpoxy Powder-coated Cold Rolled Steel
ModelPrecision SWB 15S
Temperature Presets4
Temperature Stability±0.1°C
Temperature Uniformity±0.05°C
Dimensions (L x W x H)15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
Electrical RequirementsGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Amperage9.0-10.5 / 4.3-5.0 (120V/230V)
Unit SizeEA