Improve sensitivity and detectability for analytical methods with microsphere suspensions that are internally dyed and feature bright, high-contrast colors.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Mean
Quantity
TSSR0300
3.0 μm
10 mL
TSSR0100
1.0 μm
1 Each
TSSR0200
2.0 μm
10 mL
3 Options
Catalog number TSSR0300
Price (SGD)
588.00
EA
Add to cart
Diameter (Metric) Mean:
3.0 μm
Quantity:
10 mL
Price (SGD)
588.00
EA
Add to cart
Improve sensitivity and detectability for analytical methods with Thermo Scientific™ Fluoro-Max Red Aqueous Fluorescent Particles, microsphere suspensions that are internally dyed and feature bright, high-contrast colors.
Fluoro-Max fluorescent particles emit bright and distinct colors when illuminated by the light of shorter wavelengths than the emission wavelength, improving their contrast and visibility relative to background materials.These internally dyed polymer particles utilize the exclusive Firefli™ process to incorporate the dye throughout the polymer matrix to produce bright fluorescent colors, minimize photo bleaching and prevent dye leaching into aqueous media.
Features:
Made of polystyrene (PS), which has a density of 1.05g/cm3 and a refractive index of 1.59 @ 589nm (25°C)
Aqueous suspensions are packaged as 1% solids
Can be detected with an epifluorescence microscope, confocal microscope, fluorometer, fluorescence spectrophotometer or fluorescence activated cell sorter
Can also be detected using mineral light or black light (UV)
Note:
Spectral information is approximate and for reference only. Spectral properties of the dyes are dependent on their concentration and physical environment. The exact excitation and emission maxima may vary depending on the size and composition of the particles.
Recommended for:
Filter testing
Flow tracing
Biological research
Specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene particles in water
ColorRed
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Red (542/612nm)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity10 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean3.0 μm
Diameter (Metric) Nominal3.0 μm
Diameter (Metric)3.0 μm
Unit SizeEA
Contents & Storage
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. Product comes with stir bar for continual mixing duriong sampling.