Filter
Filter
Actual product may vary
Filter
Haws™ Filter is for installation on all eyewash units for additional protection for eyes.
Have Questions?
Catalog number TSS9070
Price (SGD)
858.00
EA
Add to cart
Price (SGD)
858.00
EA
Add to cart
Model 9070 Filter is recommended for installation on all eyewash units for additional protection for eyes. Made of FDA approved polypropylene with disposable FDA approved viscose fiber filter media which removes particles down to 20 microns. Flow Capacity: 7 gpm (26.5 L) max (recommended). Maximum Working Pressure: 125 psi (8.62 bar) Size: 15 in. (38.1 cm) overall by 5 in. (12.7 cm) diameter Supply: 3/4 in. NPT inlet and outlet.
Specifications
ColorBlue, Black
Height (English)12.0625 in.
Height (Metric)30.5 cm
Product TypeFilter
Unit SizeEA