Safety Sign
Safety Sign
Actual product may vary
Safety Sign
Haws™ Safety Sign is a 3-sided high visibility sign
Have Questions?
Catalog NumberMaterial
TSS9020Plastic
Catalog number TSS9020
Price (SGD)
858.00
EA
Add to cart
Material:
Plastic
Price (SGD)
858.00
EA
Add to cart
Model 9020 ABS plastic 3-sided sign is for use on Haws combination shower and eyewash units. This large yellow ABS plastic sign with Safety Green printed stripes is visible from multiple directions. Features universal pictorial of an emergency shower and eyewash. It can be mounted on a wall for 2 way visibility, or on a shower upright for 3 sided visibility. To be used only on combination shower and eyewash units.
Specifications
Certifications/ComplianceANSI Z358.1
Electrical RequirementsNo
For Use With (Equipment)8320CRP, 8330, 8336, 8355WCC, 8355WCW, 8356WCC, 8356WCW, 8356WCDD, 8356WCSM, 8300-8309, 8300CRP-8309CRP, 8300FP, 8300.158, 8309WC, 8317, 8320-8325
MaterialPlastic
Quantity1 Ea.
Size (English)Large
Mounting TypeBracket
For Use With (Application)Emergency Showers and Eyewashes
Height (English)33.5 in.
Height (Metric)85.09 cm
Width (English)7 in.
Width (Metric)17.78 cm
Dimensions (L x W x H)7 x 7 x 33.5 in.
Length (English)7 in.
Length (Metric)17.78 cm
Shape3-sided
LanguagesUniversal
Displayed TextSafety
Product Type3-sided Safety Sign
ColorYellow
Unit SizeEA