Fluka™ Non-Polar Silica Based SPE Columns
Fluka™ Non-Polar Silica Based SPE Columns
Actual product may vary
Fluka™ Non-Polar Silica Based SPE Columns
End-capped, Medical-grade polypropylene with PE frits
Have Questions?
Catalog NumberSorbentColumn Capacity (Metric)Bed Weight
TSS9008Octadecyl (C18)6 mL1000 mg
Catalog number TSS9008
Price (SGD)
858.00
EA
Add to cart
Sorbent:
Octadecyl (C18)
Column Capacity (Metric):
6 mL
Bed Weight:
1000 mg
Price (SGD)
858.00
EA
Add to cart
  • All columns are reversed phase and should be preconditioned with a polar solvent with the elution solvent being non-polar
  • All columns are endcapped and have a surface area of 500m2/g
Specifications
Column Capacity (Metric)6 mL
Bed Weight1000 mg
Particle Size40 to 63 μm
SorbentOctadecyl (C18)
EndcappedYes
Pore Size60 Å
Quantity30
Surface Area (Metric)500 m2/g
Unit SizeEA