Check Order Status
CytoDot Pen
Actual product may vary
CytoDot Pen
Designed for ink application on glass slides and coverslips after staining.
Have Questions?
Change view
Catalog Number
Color
TSS9002
Green
9002
Green
2 Options
Catalog number
TSS9002
Price (SGD)
858.00
EA
Add to cart
Color:
Green
Price (SGD)
858.00
EA
Add to cart
Fine tip cytology markers are permanent, waterproof, and write on almost any surface including glass, plastic, ceramic and metal
4 pens per pack
Specifications
Tip Style
Fine
Color
Green
Autoclavable
Not Autoclavable
Sterility
Non-sterile
Disposable
Reusable
Latex Free
Yes