Quik-Align™ Gothic Style Replacement Ten Packs: Black 3 in. High
Quik-Align™ Gothic Style Replacement Ten Packs: Black 3 in. High
Actual product may vary
Quik-Align™ Gothic Style Replacement Ten Packs: Black 3 in. High
Quik-Align™ die-cut numbers and letters are self-spacing and self-aligning. The characters interlock to ensure perfect spacing for professional-looking results every time.
Have Questions?
Catalog NumberDisplayed Text
TSS5100AA
Catalog number TSS5100A
Price (SGD)
833.00
EA
Add to cart
Displayed Text:
A
Price (SGD)
833.00
EA
Add to cart
  • Traditional sans serif Gothic font provides excellent legibility even at a distance
  • Card-mounted vinyl film characters features a pressure sensitive adhesive
  • Good chemical, oil and water resistance
  • Average outdoor durability 2-4 years
Specifications
Certifications/ComplianceRoHS
For Use With (Equipment)General & Industrial ID
MaterialVinyl
Quantity10/Pk.
StyleGothic
Tensile Strength14 lb./in.
Adhesive TypePermanent Acrylic
For Use With (Application)Indoor and outdoor use
Height (English)3 in.
Max. Temperature (English)200°F
Min. Temperature (English)-40°F
Thickness0.005 in.
Temperature Range (Metric)-40°C to 93°C
Temperature Range (English)-40°F to 200°F
Displayed TextA
Product TypeIndividual Letter
ColorBlack on Blue
Unit SizeEA